Cell culture supplements, Panexin NTS serum substitute
Lieferant: PAN BIOTECH
Panexin NTS is a ready-to-use, fully defined serum substitute for the cultivation of suspension cells under serum-free conditions. Panexin NTS is developed with a unique technology and contains a special 3-dimensional substance release system (3D-SRS) for an optimal support of cells with nutrients and growth stimulants. This solution is added to the culture medium in a final concentration of 10%. It supports the growth of many cell types in an optimum manner.
- Sterile solution
- Store at ‒20 °C
Panexin NTS contains purified proteins, lipids, salts, amino acids, trace elements, and hormones in an optimised formulation and a new 3-dimensional substance release system (3D-SRS). Panexin NTS contains no growth factors, undefined hydrolysates or peptones.
Panexin NTS Pharma Grade is free of any animal-derived components.
Panexin NTS can be used for many cell lines to replace FBS. Due to specially selected and pretested raw materials Panexin NTS batches are very homogeneous. Therefore the complex batch testing known from FBS can be omitted with the use of Panexin NTS.
Panexin NTS is completely defined and contains no growth factors, undefined peptones or hydrolysates. Due to this special formulation the interpretation of results from studies on effects of individually added growth factors is easier and more reliable in serum-free conditions. For cell lines which require specific growth factors these should be added in a concentration as previously used.