Gentamicinsulfat 10 mg/ml
Lieferant: PAN BIOTECH
|
Achtung
|
Synonyme:
Gentamicin Sulfatsalz
P06-03050
PANBP06-03050EA
35.8
EUR
PANBP06-03050
Gentamicinsulfat 10 mg/ml
Gentamicinsulfat
Even though cells should be cultured without the addition of antibiotics or antifungal drugs, cell culture media are not only a perfect breeding ground for your cell culture but also for many contaminants, such as bacteria and fungi. In certain cases the addition of antibiotics is necessary (for unique cell lines or to remove already existing contamination).
Lagertemperatur:
Gefrierschrank |
MDL:
MFCD00270181 CAS-Nummer: 1405-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...