Calf collagen (from skin)
Lieferant: PAN BIOTECH
P06-20300
PANBP06-20300EA 864 EUR
PANBP06-20300
Calf collagen (from skin)
Proteine und Peptide
Collagen is one of the main structural components of the ECM in connective tissue and internal organs. It is mostly prevalent in skin, tendon, and bones. Type I Collagen is a heterodimer consisting of two α₁(I)-chains and one α₂(I)-chain, that spontaneously form a triple helix at neutral pH and 37 °C.
  • Culture of cells with which hardly or don’t proliferate at all on glass or plastic surfaces
  • Adherence of cells in culture media without serum or fibronectin
  • Analysis of cell migration and morphology
  • Change of cell appearance in 3D-collagen gels
  • Maintenance of Cell differentiation status of higher cells
  • Development of tissue like structures in-vitro and the application of them in wound healing processes
It is used in the production of gels, to embed cells. Type I Collagen-Gels are suitable for use as a substrate for adherent cells in a cell culture vessel or as a floating matrix in or on a cell culture medium.
Type 1 bovine calf collagen; 4 mg/ml in 0,1% acetic acid, act as a excellent substrate for the culture of epithelial cells and many other cell lines.
Bitte beachten For research use only. Not approved for human or animal diagnostic or therapeutic procedures.