Foetal bovine serum, delipidised, EU approved

Lieferant: PAN BIOTECH
P30-3402
PANBP30-3402EA 0 EUR
PANBP30-3402
Foetal bovine serum, delipidised, EU approved
Seren
In this delipidised serum, lipids are removed from serum by affinity chromatography.

  • Sterile solution
  • Store at ‒20 °C

Applications include: Lipid metabolism studies, arteriosclerosis research.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR