Digestion systems, acid digestion 'bomb' 4749
Lieferant: PARR
4749
PARR4749EA 0 EUR
PARR4749
Digestion systems, acid digestion 'bomb' 4749
Aufschlusssysteme Aufschluss-Systeme
Digestion system designed to work with strong mineral acids, organic acids and other oxidizing acids. By using this vessel samples can be dissloved or digested rapidly, for all types of process. It can use at higher temperatures and pressures, and provided with safety blow-out protection as well.
Operating temperature: max. 250 °C
Operating pressure: up to 1800 psig
Capacity: 23 ml
Bestellinformation: Accessories like spanner wrench, holding fixture and tumbling ring are not included with these vessels and must be ordered separately.