Human plasma, type 2 diabetes mellitus
Lieferant: Charles River Laboratories Cell Solutions
PB006C-1-DM2
PB006C-1-DM2EA 0 EUR
PB006C-1-DM2
Human plasma, type 2 diabetes mellitus
Plasma
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of type 2 diabetes mellitus (DM 2). Collected from IRB-consented donors diagnosed with type 2 diabetes mellitus.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Bitte beachten Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.