Human plasma, gastric cancer (GC)
Lieferant: Charles River Laboratories Cell Solutions
PB006C-1-GC
PB006C-1-GCEA 0 EUR
PB006C-1-GC
Human plasma, gastric cancer (GC)
Plasma
Plasma used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy. Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Collected from IRB-consented donors diagnosed with GC.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Bitte beachten Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.