Human plasma, multiple sclerosis (MS)
Lieferant: Charles River Laboratories Cell Solutions
PB006C-1-MS
PB006C-1-MSEA 0 EUR
PB006C-1-MS
Human plasma, multiple sclerosis (MS)
Plasma
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of multiple sclerosis (MS). Collected from IRB-consented donors diagnosed with MS.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.