Human plasma, prostate cancer
Lieferant: Charles River Laboratories Cell Solutions
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of prostate cancer (PC), the most common type of cancer in men. Collected from IRB-consented donors diagnosed with PC.
Treatment for prostate cancer is a very active area of research, where newer treatments are being developed, and improvements are being made among many standard prostate cancer treatment methods.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.