Human plasma, rheumatoid arthritis (RA)
Lieferant: Charles River Laboratories Cell Solutions
PB006C-1-RA
PB006C-1-RAEA 0 EUR
PB006C-1-RA
Human plasma, rheumatoid arthritis (RA)
Plasma
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of rheumatoid arthritis (RA). Collected from IRB-consented donors diagnosed with RA.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.