Human serum, human immunodeficiency virus (HIV)
Lieferant: Charles River Laboratories Cell Solutions
PB007C-1-HIV
PB007C-1-HIVEA 0 EUR
PB007C-1-HIV
Human serum, human immunodeficiency virus (HIV)
Seren
Human serum used for researching the causes, diagnosis, supportive care and treatment of human immunodeficiency virus (HIV). Collected from IRB-consented donors diagnosed with HIV.
There is currently no cure for HIV or AIDS, however with proper treatment we are able to keep the level of HIV virus in the body low. This donors samples can be used by researchers to investigate biomarker and to develop the diagnostic methodologies, pre-clinical research validation, and drug toxicity and efficacy.
Storage: Store at −80 °C.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.