Human mononuclear cells from peripheral blood, Hepatitis C virus (HCV)
Lieferant: Charles River Laboratories Cell Solutions
PB009C-1-HCV
PB009C-1-HCVEA 0 EUR
PB009C-1-HCV
Human mononuclear cells from peripheral blood, Hepatitis C virus (HCV)
Zellen
Mononuclear cells (MNCs) used for researching the causes, diagnosis, supportive care and treatment of Hepatitis C Virus (HCV). Human mononuclear cells are collected from human volunteer donors diagnosed with Hepatitis C virus under IRB approved informed consent.
  • Cryopreserved in CryoStor® CS10 media
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Bitte beachten Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.