Wasser Nuclease_free
Lieferant: OMEGA BIO-TEK
Synonyme:
Wasserstoffoxid, Hydrogenoxid
PD092
PD092EA
75.7
EUR
PD092
Wasser Nuclease_free
Wasser
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...