Membrane nitrogen generator, Infinity XE 60 series
Lieferant: PEAK SCIENTIFIC
3303147
PEAK3303147EA 13560 EUR
PEAK3303147
Membrane nitrogen generator, Infinity XE 60 series
Gasgeneratoren Stickstoff-Generatoren
Producing between 10 and 520 L/min of high purity nitrogen gas, the Infinity XE 60 series nitrogen membrane gas generators can comfortably supply multiple laboratory instruments with instrument grade nitrogen.
  • Flow rates ranging from 10 to 520 L/min *
  • N₂ purity up to 99,5%
  • Versatile solution with varying nitrogen flow, pressure and purity capabilities
  • In-field expansion of nitrogen capacity, for models without dry air outlet, to meet growing laboratory demands
  • Optional second outlet for dry air
  • Intelligent membrane management (IMM) optimises compressed air requirement based on gas consumption
  • IMM reduces overall compressor run time and energy consumption, reducing carbon footprint
  • Fully comprehensive 2 year warranty **
Offering laboratories a cost-effective alternative to cylinders in a compact, space saving nitrogen solution, the Infinity XE 60’s intelligent membrane Mmanagement (IMM) uniquely optimizes air consumption depending on set purity, inlet pressure/flow and nitrogen demand. With variable N₂ purities (95 to 99,5%), XE 60 can meet the demands of numerous applications such as ELSD, NMR, FT/MS, Sample Evaporators, as well as LC-MS and should your lab demands grow over time, Infinity XE 60 can have its nitrogen output increased on-site up to a maximum of 520 L/min all within the same original footprint.
To operate, the infinity series of generators require an external source of compressed air, which meets a minimum quality grade of ISO 8573-1:2010 Class 1.4.1. Where required, Peak can provide assistance on external compressors and pre/post filtration as part of delivering a complete solution for your needs.
* Dependent on inlet pressure, inlet/outlet flow and Nitrogen purity setting
** Subject to meeting manufacturers 2 year warranty conditions (see www.peakscientific.com/warranty-statement/)