Nitrogen Generator, Horizen 24
Lieferant: PEAK SCIENTIFIC
3304532
PEAK3304532EA 17476 EUR
PEAK3304532
Nitrogen Generator, Horizen 24
Gasgeneratoren Stickstoff-Generatoren
Bringing Single Quad LC-MS users the very latest in nitrogen gas generator technology, Horizen 24 is the culmination of over 25 years at the forefront of nitrogen gas generation for the laboratory.
  • The most energy-efficient nitrogen generator on the market
  • Ultra-dry, non-methane hydrocarbon-free nitrogen at up to 99,9% purity and up to 116 psi
  • 55% less energy, saving power consumption*
  • Reduction in heat output by over 50%, saving on air conditioning costs*
  • Two year comprehensive manufacturer's warranty**
  • The smallest nitrogen generator in its class, fitting easily under any laboratory bench
  • Minimised operational carbon footprint compared with cylinders and equivalent generator models
  • State-of-the-art AirMax™ technology
  • Easy to plan annual maintenance
Designed to provide a premium, yet cost-effective nitrogen solution, Horizen 24 has incorporated state-of-the-art compressor technology to efficiently manage power consumption, reducing energy costs and heat output without compromising on performance.
As a standalone solution delivering 24 litres per minute of high-purity nitrogen, up to 99,9%, from a compact footprint, Horizen 24 gives laboratories peace of mind and confidence in their LC-MS results.
* Benchmarked against equivalent competitor.
**Year 2 of warranty subject to generator being serviced at end of year 1 by a PEAK-approved agent in accordance with fixed annual maintenance schedule. For full terms and conditions visit: www.peakscientific.com/warranty-statement.