pH <791> PT, Proficiency Testing Material, Supelco®
Lieferant: Merck
PEP010-3X100ML
PEP010-3X100MLEA 219 EUR
PEP010-3X100ML
pH <791> PT, Proficiency Testing Material, Supelco®
Standards Eignungstest-Standards
Suitable for 791 per USP. Proficiency Testing Kit containing three samples ranging from pH 5,0 to 10,00.
Standards: This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. 