HPLC Columns, Brownlee™ MPLC Catridges
Lieferant: PerkinElmer
07110016
PERC07110016EA 1580 EUR
PERC07110016
HPLC Columns, Brownlee™ MPLC Catridges
Chromatographiesäulen
The MPLC cartridge system allows direct coupling of an analytical and NewGuard™ cartridge without introduction of any dead volume. A specific cartridge holder (30, 100, 220 mm) is required for each cartridge length.
  • Easy changing with fingertight cartridge
  • Avoids disconnecting of tubing from the LC system