Potassium bromide die set
Lieferant: PerkinElmer
51001144
PERC51001144EA 1010 EUR
PERC51001144
Potassium bromide die set
Pressen
The evacuable potassium bromide (KBr) die is designed to produce uniform KBr pellets.
Suitable for FT-IR spectrometry of solid samples.