Tubing assemblies, PTFE
Lieferant: PerkinElmer
B0191058 B0191059 B0191060 B0198097 B0198099 B0198100 B0501594
PERCB0191058EA 55.6 EUR
PERCB0191058 PERCB0191059 PERCB0191060 PERCB0198097 PERCB0198099 PERCB0198100 PERCB0501594
Tubing assemblies, PTFE
Schläuche Transferschlauch
Made of PTFE.
  • Very good chemical resistance
  • Very good temperature resistance