Accessories for plasma 40/plasma 400
Lieferant: PerkinElmer
N0690676
PERCN0690676EA 105 EUR
PERCN0690676
Accessories for plasma 40/plasma 400
Analysatoren Elementar-Analysatoren
This tip set includes a clear sapphire sample tip (0,3 mm Int. Ø) and a red ruby argon tip (0,22 mm Int. Ø).
The tips are made of PEEK with the gem inserts noted for maximum corrosion resistance.