Silver capsule for oxygen analysis kit
Lieferant: PerkinElmer
N2411364
PERCN2411364EA 386 EUR
PERCN2411364
Silver capsule for oxygen analysis kit
Probenbehälter Universalbehälter
Silver capsule used for analysing organic substances. For oxygen content either silver or standard tin capsules may be used for analysing organic substances for oxygen content.