Deuterium lamp for UV/Vis detector
Lieferant: PerkinElmer
N2920149
PERCN2920149EA 1140 EUR
PERCN2920149
Deuterium lamp for UV/Vis detector
Beleuchtung Deuterium-Lampen
Deuterium lamp for the LC series 200 UV/Vis detector, provide outstanding performance and lamp life.
Lamp changes are quick and easy due to a unique, self-aligning lamp mount.