Inline gas filters
Lieferant: PerkinElmer
N9301178
PERCN9301178EA 440 EUR
PERCN9301178
Inline gas filters
Filter Gasfilter
This in-line filter removes moisture, oil, and particulate from gas supplies. It consists of a molecular sieve and indicator in a clear acrylic tube. The drierite changes colour at low relative humidity indicating that packing must be changed.
  • Large capacity filter requires fewer filter changes
  • Improves baseline stability and chromatographic results
Max. pressure: 6,89 bar
Dimensions (W×H): 57×432 mm with 1/8” fittings