Multi-element calibration standard 3 without Hg
Lieferant: PerkinElmer
N9301720
PERCN9301720EA 325 EUR
PERCN9301720
Multi-element calibration standard 3 without Hg
Standards Kationischer Standards Mehrelement-Standards
10 µg/ml: Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn