Microsampling tools
Lieferant: PerkinElmer
N1870151 N9302603 N9302605 N9302609 N9302610 N9302622
PERCN1870151EA 709 EUR
PERCN1870151 PERCN9302603 PERCN9302605 PERCN9302609 PERCN9302610 PERCN9302622
Microsampling tools
Werkzeugsätze
This microsampling tool kit is suitable for FTIR microscopy.
Lieferumfang: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.