Hand crimper
Lieferant: PerkinElmer
N9302785
PERCN9302785EA 568 EUR
PERCN9302785
Hand crimper
Crimper, Capper und Decapper
Hand crimper for GC Headspace vials or LC large volume vials. It ensures sample integrity, particularly those containing volatile compounds.