Calcium Standard Solution, 1,000 μg/ml, 2% HNO₃
Lieferant: PerkinElmer
N9303733
PERCN9303733EA
80.3
EUR
PERCN9303733
Calcium Standard Solution, 1,000 μg/ml, 2% HNO₃
Standards
Kationischer Standards
Calcium-Standards
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
- Analysed by ICP-OES
- Analysed by classical wet assay
- 32 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppm level
- Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Bitte beachten These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).