Cadmium standard solution, for ICP-MS, Pure Plus grade
Lieferant: PerkinElmer
N9303734
PERCN9303734EA
89.5
EUR
PERCN9303734
Cadmium standard solution, for ICP-MS, Pure Plus grade
Standards
Kationischer Standards
Cadmium-Standards
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
- Analysed by ICP-OES
- Analysed by classical wet assay
- 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppb level
- Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Bitte beachten These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).