Calibration standard, single element, dysprosium for AAS and ICP-OES, pure grade
Lieferant: PerkinElmer
N9303768
PERCN9303768EA 57.8 EUR
PERCN9303768
Calibration standard, single element, dysprosium for AAS and ICP-OES, pure grade
Standards Kationischer Standards Dysprosium-Standards
Bitte beachten These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.