Calibration standard, single element, vanadium for AAS and ICP-OES, pure grade
Lieferant: PerkinElmer
N9303808
PERCN9303808EA 69.6 EUR
PERCN9303808
Calibration standard, single element, vanadium for AAS and ICP-OES, pure grade
Standards Kationischer Standards Vanadium-Standards
Pure Grade Standards for AA and ICP-OES
  • Analyzed by ICP-OES
  • Analyzed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Bitte beachten These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.