Calibration standard, single element, zirconium for AAS and ICP-OES, pure grade
Lieferant: PerkinElmer
N9303812
PERCN9303812EA
55.6
EUR
PERCN9303812
Calibration standard, single element, zirconium for AAS and ICP-OES, pure grade
Standards
Kationischer Standards
Zirkonium-Standards
Pure Grade standard for AA and ICP-OES.
- Analysed by ICP-OES
- Analysed by Classical Wet Assay
- 32 trace impurities Analysed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppm level
- All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Bitte beachten These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.