Digestion tubes, DigiTUBE®
Lieferant: PerkinElmer
N9308037 N9308048 N9308049 N9308050 N9308066 N9308008 N9308016 N9308024 N9308711 N9308712 N9308713
PERCN9308037EA 579 EUR
PERCN9308037 PERCN9308048 PERCN9308049 PERCN9308050 PERCN9308066 PERCN9308008 PERCN9308016 PERCN9308024 PERCN9308711 PERCN9308712 PERCN9308713
Digestion tubes, DigiTUBE®
Röhrchen Aufschlussgefäße
Disposable, sample digestion tubes with class A graduations.
Rack lock design allows for easy, one handed closure of leak free screw cap.