Shakematic, 1095
Lieferant: PerkinElmer
PER011090
PERCPER011090EA 0 EUR
PERCPER011090
Shakematic, 1095
Schüttler Rotations-/Wippschüttler
The Shakematic 1095 is designed and built specifically for mixing samples for Falling Number analysis.
  • Sample mixing time is reduced to 3 seconds which gives an increase in lab throughput
  • Operator independent results
  • Improved repeatability
  • Increased lab throughput
  • Less manual work
It has enhanced safety features, is CE-labeled and runs significantly quieter and vibration free.