Wipes for general applications, VeraClean® Critical Cleaning Plus
Lieferant: PGI NONWOVEN
74603 74606 74600 7461300
PGIN74603EA 14.7 EUR
PGIN74603 PGIN74606 PGIN74600 PGIN7461300
Wipes for general applications, VeraClean® Critical Cleaning Plus
Wischtücher Wisch- und Handtücher, Einweg Wischtücher für allgemeine Anwendungen
These semi-reusable wipes are perfectly suited for precision cleaning and critical wiping tasks across a range of industries. Thanks to their composite construction they can instantly absorb a wide range of fluids and provide a dry at once effect.
  • Durable, strong enough for multiple uses
  • High absorbency: Ideal for easy removal of fluids and spills
  • Extremely strong both wet and dry: Won't fall apart like paper products, even when used in combination with detergents, water or solvent
  • Low linting