Disinfection trays, autoclavable
Lieferant: PHAGOGENE
60275
PHAG60275EA 1940 EUR
PHAG60275
Disinfection trays, autoclavable
Schalen/Schüsseln/Töpfe Sterilisationsbehälter und -wannen
PP.
  • Polypropylene bins
  • Equipped with lid
  • Graduation inside the bins
  • Autoclavable at 134 °C/18 min.
Trays for pre-disinfection and disinfection by soaking of the medico-surgical instrumentation.