PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
Lieferant: PHAGOGENE
SA0135720
PHAGSA0135720EA 334 EUR
PHAGSA0135720
PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
Reinigungsmittel Desinfektionsmittel
Intended for the hygienic surgical disinfection of the hands by friction.
  • High tolerance
  • Speed ​​of use
  • Bactericidal, fungicidal, mycobactericidal and virucidal
  • Fragrance-free, dye-free, phenoxyethanol-free, and paraben-free
It is used as an alternative to simple or hygienic hand washing, in the absence of a water point, during serial contact, before any aseptic gesture.