Pharmaceutical secondary standard, Benzyl alcohol, (CRM), Supelco®
Lieferant: Merck
PHR1019-1G
PHR1019-1GEA
84.1
EUR
PHR1019-1G
Pharmaceutical secondary standard, Benzyl alcohol, (CRM), Supelco®
Standards
Organische Standards
- Traceable to PhEur Y0000167
- Traceable to USP 1061901
Standards: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).