4'-Ethoxyacetanilid, Supelco®

Lieferant: Merck

PHR1094-1G
PHR1094-1GEA 136 EUR
PHR1094-1G
4'-Ethoxyacetanilid, Supelco®
4'-Ethoxyacetanilid
Formel: C₁₀H₁₃NO₂
MDL: MFCD00009094
CAS-Nummer: 62-44-2
EINECS: 200-533-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30873e44ab7d5ccd1b0eed5de102befe7447a700&vid=30873e44ab7d5ccd1b0eed5de102befe7447a700&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR