Pharmaceutical secondary standard, Retinyl Acetate (Vitamin A Acetate), certified reference material, Supelco®
Lieferant: Merck
PHR1236-1G
PHR1236-1GEA 93 EUR
PHR1236-1G
Pharmaceutical secondary standard, Retinyl Acetate (Vitamin A Acetate), certified reference material, Supelco®
Standards Organische Standards
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=702aa68bed4b27ddc8256a8d5d049ef18861b8c2&vid=702aa68bed4b27ddc8256a8d5d049ef18861b8c2&prid=web&clid=SAPDAM