Organic reference standard, Inositol, CRM, Pharmaceutical Secondary Standard, Supelco®
Lieferant: Merck
PHR1351-1G
PHR1351-1GEA 96.9 EUR
PHR1351-1G
Organic reference standard, Inositol, CRM, Pharmaceutical Secondary Standard, Supelco®
Standards Organische Standards
A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.
  • Vapour density: 6,2 (vs air)
  • Melting point: 222 to 227 °C
These pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28330674f7e11310d138f8f825e41b970a20107a&vid=28330674f7e11310d138f8f825e41b970a20107a&prid=web&clid=SAPDAM