Organic reference standard, Inositol, CRM, Pharmaceutical Secondary Standard, Supelco®
Lieferant: Merck
PHR1351-1G
PHR1351-1GEA
96.9
EUR
PHR1351-1G
Organic reference standard, Inositol, CRM, Pharmaceutical Secondary Standard, Supelco®
Standards
Organische Standards
A component of membrane phospholipids, glycosylphosphatidylinositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.
- Vapour density: 6,2 (vs air)
- Melting point: 222 to 227 °C
These pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.