Pharmaceutical secondary standard, Fluoxetine Hydrochloride, certified reference material, Supelco®
Lieferant: Merck
PHR1394-1G
PHR1394-1GEA
100
EUR
PHR1394-1G
Pharmaceutical secondary standard, Fluoxetine Hydrochloride, certified reference material, Supelco®
Standards
Organische Standards
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.