Pharmaceutical secondary standard, Quercetin, certified reference material, Supelco®
Lieferant: Merck
PHR1488-1G
PHR1488-1GEA 106 EUR
PHR1488-1G
Pharmaceutical secondary standard, Quercetin, certified reference material, Supelco®
Standards Organische Standards
Traceable to Ph. Eur. Y0001009 and to USP 1592409.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad2c427829be16cc636a85f2560b1f47cb4f0cc&vid=1ad2c427829be16cc636a85f2560b1f47cb4f0cc&prid=web&clid=SAPDAM