Pharmaceutical secondary standard, lidocaine impiurity G, (CRM), Supelco®

Lieferant: Merck

PHR1995-50MG
PHR1995-50MGEA 627 EUR
PHR1995-50MG
Pharmaceutical secondary standard, lidocaine impiurity G, (CRM), Supelco®
Standards Organische Standards

Bestellen

TECHNISCHE DATEN

Bezeichnung Lidocaine Impurity G
Environmentally Preferable
CAS Nr. 35891-87-3
Formel C₁₃H₂₀N₂O · HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR