Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Lieferant: Thermo Fisher Scientific
15031
PIER15031EA 848 EUR
PIER15031
Pierce™ Polystyrene Strip Plates, 8-Well, Corner Notch
Mikrotiterplatten Immunologie-Mikrotiterplatten
PS, high antibody-binding 96-well microplate with individual 8-well strips in the plate frame for immunoassay applications.
  • Clear polystyrene for colorimetric detections
  • Plates are certified for performance, having consistently low coefficients of variation among wells
  • Hydrophobic-ionic binding interaction
  • Wells can be easily separated from strips with gentle twist
  • Corner notch for use in standard plate readers