Glutathione spin columns, Pierce™
Lieferant: Thermo Fisher Scientific
16105 16103 16104
PIER16105EA 506 EUR
PIER16105 PIER16103 PIER16104
Glutathione spin columns, Pierce™
Filter Zentrifugaleinheiten
These spin columns provide a fast, easy to use format for purifying GST-fusion proteins from cellular lysates. The glutathione is immobilised through its central sulfhydryl onto 6% crosslinked agarose resin. Purification of GST-fusion proteins using glutathione (GSH) agarose beads is well documented and provides a one-step, high purity affinity purification.
  • Binds at least 40 mg of purified recombinant GST protein per ml of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per ml of resin with greater than 90% purity
  • Resin can be reused at
More Product Information