Desthiobiotin NHS ester, EZ-Link™, Pierce™

Lieferant: Thermo Fisher Scientific

16129
PIER16129EA 294 EUR
PIER16129
Desthiobiotin NHS ester, EZ-Link™, Pierce™
Desthiobiotin NHS ester

Thermo Scientific EZ-Link NHS-Desthiobiotin is a short, membrane permeable, amine-reactive labeling reagent whose biotin-like group is elutable from streptavidin, making it ideal for intracellular protein labeling and purification.


  • Desthiobiotin – analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
  • Protein labeling – tag antibodies or proteins in purified or mixed samples to enable high recovery in pull-down assays with streptavidin beads
  • Membrane-permeable – achieve intracellular labeling with intact cells because the small, uncharged reagent easily diffuses across cell membranes
  • Amine-reactive – reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides


NHS-Desthiobiotin is a variant of biotin that is activated as an NHS ester to covalently label primary amines (-NH2) of proteins or other molecules with desthiobiotin groups. The desthiobiotin tag binds to streptavidin and other biotin-binding proteins with high specificity yet readily elutes with mild conditions (i.e., by competitive displacement with regular, free biotin). As such, this reagent is a useful alternative to NHS-Biotin (Part No. 20217) for avidin-biotin techniques in which nondenaturing elution of the labeled proteins is desired. This simplest and smallest variety of amine-reactive desthiobiotin reagent is uncharged and cell membrane permeable; therefore, NHS-Desthiobiotin can be used with intact cells for intracellular protein labeling.


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₄H₂₁N₅O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD08062086
CAS-Nummer: 80750-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR