Desthiobiotinylation and pull-down kit, EZ-Link™
Lieferant: Thermo Fisher Scientific
16138
PIER16138EA 702 EUR
PIER16138
Desthiobiotinylation and pull-down kit, EZ-Link™
Proteinkonjugate und Markierungen
EZ-Link™ Desthiobiotinylation and Pull-down Kit is a complete set of reagents to biotin-label purified proteins and use them to purify interaction complexes using streptavidin agarose resin with mild elution conditions.
  • Sulpho-NHS-LC-Desthiobiotin label proteins with a biotin-like that specifically binds streptavidin
  • Simple, optimised protocol and reagents to label proteins
  • Recover (purify) intact, functional protein interaction complexes
  • PEG groups in spacer arms enhance water solubility of biotinylated molecules and help prevent aggregation of biotinylated antibodies
EZ-Link™ Desthiobiotinylation and Pull-down Kit uses Sulpho-NHSLC-Desthiobiotin reagents to label target proteins on primary amines, desalting columns to clean up the labeling reaction, and immobilized high capacity streptavidin resin and buffers to perform pull-down assays. Pull-down assays use the tagged or labelled bait protein coupled to resin to capture a prey protein contained in a cell lysate or other unpurified protein mixtures. This technique can identify novel interactions between a known protein (bait) and previously undiscovered target (prey) or it can confirm the interaction between a known (bait) and a known target (prey) protein.
EZ-Link™ Hydrazide-PEG4-Desthiobiotin, Amine-PEG4-Desthiobiotin, Phosphine-PEG4-Desthiobiotin and Sulpho-NHS-LC-Desthiobiotin reagents are available packaged in a convenient No-Weigh™ format in screw-cap vials, eliminating difficulties associated with weighing small quantities of reagent and enabling resealing of the vial, which reduces the risk of hydrolysis.
Lieferumfang: EZ-Link™ Desthiobiotinylation and Pull-down Kit includes EZ-Link Sulpho-NHS-LC-Desthiobiotin (2×1 mg), Zeba™ spin desalting column, 7K MWCO, 5 ml (5 columns), high capacity streptavidin agarose resin (2 ml), PBS (1 pack, makes 500 ml), Biotin elution buffer (8 ml).