Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
17926
PIER17926EA
162
EUR
PIER17926
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel:
NH₂C(CH₂OH)₃ Siedepunkt: 357 °C (1013 hPa) Schmelzpunkt: 169…172 °C Dichte: 1,328 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004679 CAS-Nummer: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...