Brij® 35 30% in wässriger Lösung, Pierce™

Lieferant: Thermo Fisher Scientific

Synonyme: Brij® L23

20150
PIER20150EA 226 EUR
PIER20150
Brij® 35 30% in wässriger Lösung, Pierce™
Brij® 35

This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 30% (w/v) solution of Brij-35 detergent for use in various protein methods.


  • A nonionic detergent for use in various protein methods
  • 30% detergent solution in ultrapure water
  • Clear, colorless viscous liquid, free of foreign material


Brij™ 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.


Detergent Class: Nonionic
Aggregation Number: 40
Micelle Molecular Weight: 49,000g
Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
Cloud Point:>100˚C
Dialyzable: No


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₅₈H₁₁₈O₂₄
Molecular Weight: 1199.56 g/mol
MDL: MFCD00080891
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR