Coupling resin, CarboxyLink™
Lieferant: Thermo Fisher Scientific
20266
PIER20266EA
267
EUR
PIER20266
Coupling resin, CarboxyLink™
Protein-Aufbereitungs
Protein Purification Resins
CarboxyLink™ coupling resin is used for covalent immobilisation of peptides or other carboxyl-containing molecules to a porous, beaded resin for use in affinity purification procedures.
- DADPA-activated crosslinked 4% beaded agarose
- Couple 1 to 2 mg of peptide per ml of resin
- Immobilises peptides with high capacity and various orientations without steric hindrance
- Immoblisation results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin